siRNA Details
Gene Symbol: RGS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002928.3 | NP_002919.3 | 4 | 500 | ||
GenBank mRNA | AF493937.1 | - | 4 | 346 | ||
AK301261.1 | - | 4 | 476 | |||
AK311880.1 | - | 4 | 476 | |||
AW193671.1 | - | 4 | 495 | |||
BC006243.2 | - | 4 | 462 | 3946461 | ||
BT006638.1 | - | 4 | 346 | |||
DA060276.1 | - | 1 | 218 | |||
DC349953.1 | - | 4 | 501 | |||
U70426.1 | - | 4 | 438 | |||
U94829.1 | - | 4 | 349 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
