siRNA Details
Gene Symbol: C1orf123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304759.1 | NP_001291688.1 | 7 | 862 | ||
NM_001304760.1 | NP_001291689.1 | 6 | 778 | |||
NM_017887.2 | NP_060357.1 | 8 | 919 | |||
GenBank mRNA | AK000587.1 | - | 8 | 897 | ||
AK092734.1 | - | 6 | 1763 | |||
BC010908.1 | - | 8 | 901 | 4093763 |
