siRNA Details
Gene Symbol: ARHGEF10L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018125.3 | NP_060595.3 | 8 | 875 | ||
NR_137287.1 | 8 | 878 | ||||
XM_017001621.1 | XP_016857110.1 | 7 | 858 | |||
XM_017001622.1 | XP_016857111.1 | 7 | 858 | |||
GenBank mRNA | AB046846.1 | - | 8 | 875 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
