siRNA Details
Gene Symbol: KIAA1551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018169.3 | NP_060639.3 | 4 | 4976 | ||
XM_005253405.2 | XP_005253462.1 | 3 | 5427 | |||
XM_006719099.2 | XP_006719162.1 | 4 | 5595 | |||
XM_011520722.1 | XP_011519024.1 | 4 | 5136 | |||
XM_017019550.1 | XP_016875039.1 | 3 | 5836 | |||
GenBank mRNA | AK001514.1 | - | 1 | 460 | ||
BC098115.1 | - | 1 | 440 | |||
BC114509.1 | - | 1 | 440 | |||
BC114956.1 | - | 1 | 440 | |||
BC140819.1 | - | 4 | 4927 | 9021687 |
