siRNA Details
Gene Symbol: C1orf106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142569.2 | NP_001136041.1 | 10 | 2665 | ||
NM_018265.3 | NP_060735.3 | 10 | 2601 | |||
XM_011509754.2 | XP_011508056.1 | 10 | 3380 | |||
XM_011509755.1 | XP_011508057.1 | 10 | 2642 | |||
GenBank mRNA | AK001583.1 | - | 3 | 1246 | ||
AK001763.1 | - | 10 | 2559 | |||
AK025066.1 | - | 1 | 420 |
