siRNA Details
Gene Symbol: S100A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002963.3 | NP_002954.2 | 3 | 265 | ||
GenBank mRNA | BC034687.1 | - | 3 | 233 | 4751685 | |
BF090388.1 | - | 3 | 316 | |||
BX476368.1 | - | 2 | 226 | |||
CR542164.1 | - | 2 | 194 | |||
M86757.1 | - | 3 | 243 |
Gene Symbol: S100A7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176823.3 | NP_789793.1 | 3 | 251 | ||
GenBank mRNA | AY189118.1 | - | 3 | 251 | ||
AY189119.1 | - | 2 | 333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
