siRNA Details
Gene Symbol: TMIGD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081976.2 | NP_001075445.1 | 3 | 566 | ||
NM_001302680.1 | NP_001289609.1 | 2 | 302 | |||
NM_020683.6 | NP_065734.5 | 3 | 1525 | |||
GenBank mRNA | AY358644.1 | - | 3 | 892 | ||
BC041707.1 | - | 3 | 504 | 5744367 | ||
BC064411.1 | - | 3 | 504 | 5744223 | ||
BI828255.1 | - | 2 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
