siRNA Details
Gene Symbol: KIAA1324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267048.1 | NP_001253977.1 | 3 | 541 | ||
NM_001284352.1 | NP_001271281.1 | 2 | 420 | |||
NM_020775.4 | NP_065826.2 | 3 | 541 | |||
XM_011541825.1 | XP_011540127.1 | 3 | 401 | |||
XM_011541826.2 | XP_011540128.1 | 3 | 401 | |||
XM_011541827.1 | XP_011540129.1 | 3 | 401 | |||
XM_017001883.1 | XP_016857372.1 | 3 | 488 | |||
GenBank mRNA | AK057647.1 | - | 3 | 399 | ||
AK297221.1 | - | 2 | 214 | |||
AK297478.1 | - | 3 | 335 | |||
AK304003.1 | - | 2 | 210 | |||
AY358366.1 | - | 4 | 457 | |||
BC125208.1 | - | 3 | 349 | 40123480 | ||
CA775143.1 | - | 3 | 367 | |||
CB142078.1 | - | 3 | 396 |
