siRNA Details
Gene Symbol: KIAA1324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267048.1 | NP_001253977.1 | 3 | 590 | ||
NM_001284352.1 | NP_001271281.1 | 2 | 469 | |||
NM_020775.4 | NP_065826.2 | 3 | 590 | |||
XM_011541825.1 | XP_011540127.1 | 3 | 450 | |||
XM_011541826.2 | XP_011540128.1 | 3 | 450 | |||
XM_011541827.1 | XP_011540129.1 | 3 | 450 | |||
XM_017001883.1 | XP_016857372.1 | 3 | 537 | |||
GenBank mRNA | AK057647.1 | - | 3 | 448 | ||
AK297221.1 | - | 2 | 263 | |||
AK297478.1 | - | 3 | 384 | |||
AK304003.1 | - | 2 | 259 | |||
AY358366.1 | - | 4 | 506 | |||
BC125208.1 | - | 3 | 398 | 40123480 | ||
CA775143.1 | - | 3 | 416 | |||
CB142078.1 | - | 3 | 445 |
