siRNA Details
Gene Symbol: PBLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022129.3 | NP_071412.2 | 10 | 1617 | ||
XM_005270028.3 | XP_005270085.1 | 10 | 1651 | |||
XM_011540060.2 | XP_011538362.1 | 10 | 1626 | |||
XM_017016513.1 | XP_016872002.1 | 10 | 1585 | |||
GenBank mRNA | AB049758.1 | - | 8 | 1480 | ||
AK027673.1 | - | 10 | 1584 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
