siRNA Details
Gene Symbol: C11orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003676.2 | NP_001003676.1 | 8 | 1868 | ||
NM_001003677.2 | NP_001003677.1 | 9 | 1613 | |||
NM_001278222.1 | NP_001265151.1 | 8 | 1492 | |||
NM_024113.4 | NP_077018.1 | 9 | 1595 | |||
NR_103471.1 | 10 | 1840 | ||||
NR_103472.1 | 8 | 1464 | ||||
XM_006718315.1 | XP_006718378.1 | 9 | 1052 | |||
XM_011520364.2 | XP_011518666.1 | 8 | 1675 | |||
XM_011520365.2 | XP_011518667.1 | 8 | 3353 | |||
XM_017018277.1 | XP_016873766.1 | 8 | 1423 | |||
XM_017018278.1 | XP_016873767.1 | 10 | 1787 | |||
XM_017018279.1 | XP_016873768.1 | 10 | 1850 | |||
XM_017018280.1 | XP_016873769.1 | 11 | 2511 | |||
XM_017018281.1 | XP_016873770.1 | 10 | 2480 | |||
XM_017018282.1 | XP_016873771.1 | 10 | 1769 | |||
GenBank mRNA | AK025863.1 | - | 9 | 1504 | ||
AK124296.1 | - | 1 | 2021 | |||
AK300813.1 | - | 8 | 1402 | |||
AL136575.1 | - | 8 | 1787 | |||
BC001860.2 | - | 9 | 1541 | 3534541 | ||
BC009455.1 | - | 8 | 1789 | 3535652 | ||
BC013988.2 | - | 9 | 1524 | 3349493 | ||
BC052291.1 | - | 8 | 1781 | 5936112 | ||
BI763384.1 | - | 1 | 429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
