siRNA Details
Gene Symbol: NECTIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002855.4 | NP_002846.3 | 5, 6 | 1170 | ||
GenBank mRNA | AF060231.1 | - | 5, 6 | 1154 | ||
BC104948.1 | - | 5, 6 | 1076 | 8143951 | ||
BC113471.1 | - | 5, 6 | 1072 | 8322523 | ||
X76400.2 | - | 5, 6 | 989 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
