siRNA Details
Gene Symbol: KIAA0319L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024874.4 | NP_079150.3 | 19 | 3052 | ||
XM_006710907.1 | XP_006710970.1 | 19 | 2851 | |||
XM_006710909.1 | XP_006710972.1 | 19 | 2943 | |||
XM_006710910.1 | XP_006710973.1 | 18 | 2495 | |||
XM_011542179.2 | XP_011540481.1 | 19 | 3183 | |||
XM_017002366.1 | XP_016857855.1 | 19 | 3036 | |||
XM_017002367.1 | XP_016857856.1 | 19 | 3198 | |||
XM_017002368.1 | XP_016857857.1 | 19 | 2866 | |||
XM_017002369.1 | XP_016857858.1 | 19 | 2958 | |||
XM_017002374.1 | XP_016857863.1 | 18 | 2510 | |||
XM_017002375.1 | XP_016857864.1 | 16 | 2152 | |||
XM_017002376.1 | XP_016857865.1 | 16 | 2137 | |||
XM_017002378.1 | XP_016857867.1 | 10 | 1176 | |||
GenBank mRNA | AB058740.1 | - | 14 | 1750 | ||
AF275679.2 | - | 19 | 3001 | |||
AF289597.1 | - | 17 | 2617 | |||
AK024287.1 | - | 15 | 1815 | |||
AK090878.1 | - | 10 | 1177 | |||
AK314605.1 | - | 19 | 3000 | |||
AL834315.1 | - | 15 | 1886 | |||
AY163234.1 | - | 19 | 3001 | |||
BC001858.2 | - | 3 | 214 | |||
BC007645.1 | - | 6 | 706 | |||
BC014530.1 | - | 19 | 2984 | 3843301 | ||
BC031672.1 | - | 14 | 1746 |
