siRNA Details
Gene Symbol: STK40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282546.1 | NP_001269475.1 | 4, 5 | 581 | ||
NM_001282547.1 | NP_001269476.1 | 4, 5 | 566 | |||
NM_032017.2 | NP_114406.1 | 5, 6 | 803 | |||
GenBank mRNA | AK024504.1 | - | 3, 4 | 365 | ||
AK075029.1 | - | 1, 2 | 82 | |||
AK075048.1 | - | 4, 5 | 563 | |||
AL834137.1 | - | 5, 6 | 760 | |||
AY198395.1 | - | 4, 5 | 536 | |||
BC007835.2 | - | 4, 5 | 534 | 4109652 | ||
BC008344.1 | - | 4, 5 | 542 | 3530483 |
