siRNA Details
Gene Symbol: FAM160A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098794.1 | NP_001092264.1 | 9 | 2311 | ||
NM_032127.3 | NP_115503.2 | 9 | 2353 | |||
XM_006718343.2 | XP_006718406.1 | 9 | 2275 | |||
XM_011520397.2 | XP_011518699.1 | 9 | 2289 | |||
XM_011520398.2 | XP_011518700.1 | 9 | 2334 | |||
XM_011520399.2 | XP_011518701.1 | 9 | 2334 | |||
GenBank mRNA | AB051546.1 | - | 9 | 2216 | ||
AK056468.1 | - | 9 | 2263 | |||
AK291702.1 | - | 9 | 2236 | |||
AL136723.1 | - | 9 | 2287 |
