siRNA Details
Gene Symbol: FCRLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184866.1 | NP_001171795.1 | 5 | 885 | ||
NM_001184867.1 | NP_001171796.1 | 3 | 600 | |||
NM_001184870.1 | NP_001171799.1 | 3 | 465 | |||
NM_001184872.1 | NP_001171801.1 | 2 | 447 | |||
NM_032738.3 | NP_116127.3 | 4 | 867 | |||
XM_006711581.3 | XP_006711644.1 | 4 | 568 | |||
XM_011510064.1 | XP_011508366.1 | 5 | 625 | |||
GenBank mRNA | AF329489.1 | - | 4 | 590 | ||
AF329491.1 | - | 2 | 210 | |||
AF329493.1 | - | 3 | 307 | |||
AF329494.1 | - | 3 | 172 | |||
AF426461.1 | - | 4 | 635 | |||
AF531423.1 | - | 4 | 867 | |||
AY091642.2 | - | 5 | 592 | |||
AY358348.1 | - | 4 | 582 | |||
BC006521.2 | - | 4 | 682 | 3345729 | ||
BX649184.1 | - | 2 | 1425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
