siRNA Details
Gene Symbol: IZUMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK294083.1 | - | 7 | 691 | ||
AY358444.1 | - | 7 | 1207 | |||
BC010443.1 | - | 6 | 918 | |||
BC040964.1 | - | 6 | 920 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
