siRNA Details
Gene Symbol: CCDC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080654.2 | NP_542385.1 | 3 | 1233 | ||
GenBank mRNA | BC008496.1 | - | 3 | 1201 | 4276656 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
