siRNA Details
Gene Symbol: TSPAN18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130783.4 | NP_570139.3 | 9 | 2587 | ||
XM_005253217.3 | XP_005253274.1 | 11 | 2771 | |||
XM_006718372.3 | XP_006718435.1 | 11 | 2858 | |||
XM_006718373.3 | XP_006718436.1 | 10 | 2844 | |||
XM_011520459.2 | XP_011518761.1 | 10 | 2734 | |||
XM_017018530.1 | XP_016874019.1 | 10 | 2609 | |||
XM_017018531.1 | XP_016874020.1 | 9 | 2531 | |||
GenBank mRNA | AK027515.1 | - | 1 | 769 | ||
AK027715.1 | - | 8 | 2396 | |||
AK075547.1 | - | 7 | 2320 | |||
AY358087.1 | - | 8 | 2510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
