siRNA Details
Gene Symbol: RFC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002915.3 | NP_002906.1 | 9 | 1568 | ||
GenBank mRNA | BC000149.2 | - | 9 | 1499 | 2900958 | |
L07541.1 | - | 9 | 1467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
