siRNA Details
Gene Symbol: RGS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002926.3 | NP_002917.1 | 16, 17 | 4476 | ||
NM_198227.1 | NP_937870.1 | 14, 15 | 1899 | |||
NM_198229.2 | NP_937872.1 | 16, 17 | 4476 | |||
XM_006713905.1 | XP_006713968.1 | 16, 17 | 3795 | |||
XM_006713907.1 | XP_006713970.1 | 14, 15 | 1853 | |||
XM_011513543.2 | XP_011511845.1 | 15, 16 | 1751 | |||
XM_017008529.1 | XP_016864018.1 | 16, 17 | 5045 | |||
XM_017008531.1 | XP_016864020.1 | 14, 15 | 2970 | |||
XM_017008532.1 | XP_016864021.1 | 13, 14 | 2593 | |||
XM_017008533.1 | XP_016864022.1 | 14, 15 | 1704 | |||
GenBank mRNA | AF030109.1 | - | 14, 15 | 1899 | ||
AF030110.1 | - | 13, 14 | 1598 | |||
AF030111.1 | - | 16, 17 | 4475 | |||
AF030112.1 | - | 12, 13 | 2529 | |||
AF035152.1 | - | 15, 16 | 3626 | |||
AY987042.1 | - | 13, 14 | 1577 | |||
BM718427.1 | - | 1, 2 | 73 | |||
CB242608.1 | - | 1, 2 | 107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
