siRNA Details
Gene Symbol: DDIAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145018.3 | NP_659455.3 | 6 | 2138 | ||
XM_011544835.1 | XP_011543137.1 | 5 | 2000 | |||
XM_011544836.2 | XP_011543138.1 | 5 | 1955 | |||
XM_011544837.1 | XP_011543139.1 | 4 | 1850 | |||
XM_011544838.2 | XP_011543140.1 | 5 | 1984 | |||
XM_017017352.1 | XP_016872841.1 | 4 | 1855 | |||
XM_017017353.1 | XP_016872842.1 | 4 | 1822 | |||
GenBank mRNA | AK023998.1 | - | 1 | 1423 | ||
AK292399.1 | - | 6 | 2097 | |||
AK297366.1 | - | 4 | 1823 | |||
BC039268.1 | - | 6 | 2106 | 5296465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
