siRNA Details
Gene Symbol: KLHDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007255.2 | NP_001007256.1 | 1 | 238 | ||
NM_152366.4 | NP_689579.3 | 1 | 238 | |||
GenBank mRNA | AK093268.1 | - | 1 | 227 | ||
AY601914.1 | - | 1 | 237 | |||
BC022077.1 | - | 1 | 237 | 4822843 | ||
BC032482.1 | - | 1 | 256 | 5222141 | ||
BC066896.1 | - | 1 | 232 | 4828667 | ||
BG718418.1 | - | 1 | 236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
