siRNA Details
Gene Symbol: CNIH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322302.1 | NP_001309231.1 | 7 | 1442 | ||
NM_001322303.1 | NP_001309232.1 | 6 | 1376 | |||
NM_001322304.1 | NP_001309233.1 | 7 | 1520 | |||
NM_001322305.1 | NP_001309234.1 | 5 | 1814 | |||
NM_152495.1 | NP_689708.1 | 6 | 1862 | |||
NR_136287.1 | 5 | 1749 | ||||
NR_136288.1 | 7 | 1372 | ||||
NR_136289.1 | 8 | 1508 | ||||
NR_136290.1 | 7 | 1442 | ||||
NR_136291.1 | 8 | 1578 | ||||
NR_136292.1 | 7 | 1512 | ||||
NR_136293.1 | 10 | 1748 | ||||
NR_136294.1 | 9 | 1682 | ||||
NR_136295.1 | 8 | 1553 | ||||
NR_136296.1 | 8 | 1578 | ||||
NR_136297.1 | 7 | 1442 | ||||
GenBank mRNA | AF070524.1 | - | 5 | 1068 | ||
AK096312.1 | - | 6 | 1862 | |||
BC022780.1 | - | 6 | 1349 | 3894832 |
