siRNA Details
Gene Symbol: TBCEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130047.1 | NP_001123519.1 | 8 | 3800 | ||
NM_152715.3 | NP_689928.3 | 8 | 3927 | |||
XM_017017326.1 | XP_016872815.1 | 9 | 4026 | |||
GenBank mRNA | AK095817.1 | - | 1 | 381 | ||
BC020501.1 | - | 8 | 3836 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
