siRNA Details
Gene Symbol: TIPRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152902.4 | NP_690866.1 | 6 | 864 | ||
GenBank mRNA | AB097034.1 | - | 6 | 747 | ||
AK313520.1 | - | 6 | 864 | |||
AL049670.1 | - | 6 | 776 | |||
BX648646.1 | - | 1 | 549 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
