siRNA Details
Gene Symbol: OVOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL831947.1 | - | 3045 | |||
BC046190.1 | - | 6 | 889 | |||
CR749395.1 | - | 5 | 3545 |
Gene Symbol: OVOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647938.1 | - | 12 | 7579 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
