siRNA Details
Gene Symbol: GOSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007024.1 | NP_001007025.1 | 9 | 2465 | ||
NM_001007025.1 | NP_001007026.1 | 9 | 2443 | |||
NM_004871.2 | NP_004862.1 | 9 | 2449 | |||
XM_005258070.4 | XP_005258127.1 | 9 | 2690 | |||
XM_005258072.2 | XP_005258129.1 | 9 | 2477 | |||
XM_017025374.1 | XP_016880863.1 | 8 | 2663 | |||
XM_017025375.1 | XP_016880864.1 | 8 | 2422 | |||
XM_017025376.1 | XP_016880865.1 | 8 | 2404 | |||
GenBank mRNA | BC040072.2 | - | 1 | 219 | ||
BC040471.1 | - | 9 | 2430 | 4697067 | ||
CB054624.1 | - | 1 | 68 |
Gene Symbol: ANKRD20A9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027995.1 | 1 | 36022 |
Gene Symbol: C11orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130935.1 | 3 | 1252 | |||
GenBank mRNA | AK075315.1 | - | 3 | 1252 |
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308043.1 | NP_001294972.1 | 5 | 4105 | ||
GenBank mRNA | AB209600.1 | - | 5 | 3802 |
Gene Symbol: DHTKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL359587.2 | - | 4 | 3500 |
Gene Symbol: ERVK-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC092439.1 | - | 4 | 672 |
Gene Symbol: HLA-DQB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097297.1 | - | 1 | 1677 |
Gene Symbol: LEPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003680.3 | NP_001003680.1 | 20 | 2902 | ||
NM_001198687.1 | NP_001185616.1 | 19 | 2867 | |||
GenBank mRNA | U52912.1 | - | 19 | 2813 | ||
U66496.1 | - | 20 | 2910 |
Gene Symbol: LOC100506634 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF469043.1 | - | 2 | 2698 |
Gene Symbol: LOC102724699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125409.1 | 4 | 1659 | |||
GenBank mRNA | AK094454.1 | - | 4 | 1659 |
Gene Symbol: LOC105373738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024142.1 | - | 2 | 1610 |
Gene Symbol: LOC105376177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056351.1 | - | 4 | 2161 |
Gene Symbol: SLC38A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128743.1 | - | 2971 |
Gene Symbol: TDRKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK225160.1 | - | 15 | 3534 |
Gene Symbol: TRIM56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516589.2 | XP_011514891.1 | 2 | 6004 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
