siRNA Details
Gene Symbol: LINC01555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027379.1 | 3 | 251 | |||
GenBank mRNA | AK092806.1 | - | 3 | 251 | ||
BC137361.1 | - | 2 | 148 | |||
BC137362.1 | - | 2 | 148 | |||
BC144607.1 | - | 3 | 196 | |||
BC171792.1 | - | 3 | 196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
