siRNA Details
Gene Symbol: CSNK2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001895.3 | NP_001886.1 | 13 | 1946 | ||
NM_177559.2 | NP_808227.1 | 14 | 2063 | |||
NM_177560.2 | NP_808228.1 | 12 | 1736 | |||
GenBank mRNA | BC050036.1 | - | 13 | 1835 | 5298655 | |
BC053532.1 | - | 12 | 1765 | 3908058 | ||
BC071167.1 | - | 12 | 1677 | 5772615 | ||
J02853.1 | - | 13 | 1822 | |||
M55265.1 | - | 13 | 1818 | |||
S53149.1 | - | 13 | 1822 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
