siRNA Details
Gene Symbol: RAB7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164522.2 | NP_001157994.1 | 6 | 1314 | ||
NM_001304839.1 | NP_001291768.1 | 5 | 1188 | |||
NM_177403.5 | NP_796377.3 | 5 | 1388 | |||
GenBank mRNA | AK122657.1 | - | 5 | 1183 | ||
AK127140.1 | - | 5 | 1372 | |||
AY094596.1 | - | 6 | 1261 | |||
BC017092.1 | - | 6 | 1256 | 3851998 |
