siRNA Details
Gene Symbol: LINC01089 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002809.2 | 7 | 1412 | |||
GenBank mRNA | AK094482.1 | - | 1 | 1504 | ||
AK095700.1 | - | 1 | 2454 | |||
BC038786.1 | - | 7 | 1412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
