siRNA Details
Gene Symbol: MTMR9LP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026850.1 | 7 | 2248 | |||
GenBank mRNA | AK055560.1 | - | 8 | 1980 | ||
AK096109.1 | - | 6 | 1800 | |||
AL833686.1 | - | 9 | 2548 | |||
BC016171.1 | - | 4 | 1093 | |||
BC041632.1 | - | 4 | 903 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
