siRNA Details
Gene Symbol: LINC00303 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027902.1 | 6 | 1261 | |||
GenBank mRNA | AK097662.1 | - | 5 | 1638 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00303 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027902.1 | 6 | 1261 | |||
GenBank mRNA | AK097662.1 | - | 5 | 1638 |