siRNA Details
Gene Symbol: DVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004423.3 | NP_004414.3 | 15 | 2739 | ||
XM_011512513.2 | XP_011510815.1 | 12 | 2527 | |||
GenBank mRNA | D86963.1 | - | 15 | 2739 | ||
U75651.1 | - | 15 | 2992 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
