siRNA Details
Gene Symbol: MXRA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005257382.2 | XP_005257439.1 | 1 | 290 | ||
XM_005257383.2 | XP_005257440.1 | 1 | 290 | |||
XM_005257384.2 | XP_005257441.1 | 1 | 290 | |||
GenBank mRNA | AK128460.1 | - | 1 | 1847 | ||
AY426672.1 | - | 1 | 274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
