siRNA Details
Gene Symbol: MXRA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008529.2 | NP_001008529.1 | 5 | 1064 | ||
NM_198530.3 | NP_940932.2 | 4 | 983 | |||
NR_130926.1 | 4 | 682 | ||||
NR_130927.1 | 5 | 881 | ||||
NR_130928.1 | 4 | 800 | ||||
XM_005257383.2 | XP_005257440.1 | 4 | 1150 | |||
XM_005257384.2 | XP_005257441.1 | 3 | 1069 | |||
GenBank mRNA | AK127801.1 | - | 1 | 2544 | ||
AK128460.1 | - | 3 | 2626 | |||
AW888225.1 | - | 5 | 799 | |||
BC053983.1 | - | 1 | 983 | 5470630 | ||
BC094776.1 | - | 1 | 954 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
