siRNA Details
Gene Symbol: FAM111B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142703.1 | NP_001136175.1 | 3 | 379 | ||
NM_001142704.1 | NP_001136176.1 | 2 | 334 | |||
NM_198947.3 | NP_945185.1 | 4 | 546 | |||
GenBank mRNA | AK303520.1 | - | 3 | 328 | ||
AK304258.1 | - | 4 | 494 | |||
AY457926.1 | - | 4 | 495 | |||
BC005998.1 | - | 4 | 496 | |||
BC130513.1 | - | 3 | 436 | 40146544 | ||
BC130539.1 | - | 3 | 436 | 40146570 | ||
DC353500.1 | - | 2 | 282 |
