siRNA Details
Gene Symbol: SLC22A25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091990.1 | - | 9 | 1701 | ||
AK127610.1 | - | 7 | 1868 | |||
AY517500.1 | - | 9 | 1701 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
