siRNA Details
Gene Symbol: IGF2BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291869.1 | NP_001278798.1 | 9, 10 | 1176 | ||
NM_001291872.1 | NP_001278801.1 | 8, 9 | 978 | |||
NM_001291873.1 | NP_001278802.1 | 8, 9 | 960 | |||
NM_001291875.1 | NP_001278804.1 | 7, 8 | 896 | |||
NM_006548.4 | NP_006539.3 | 9, 10 | 1158 | |||
XM_006713477.3 | XP_006713540.1 | 4, 5 | 582 | |||
XM_017005557.1 | XP_016861046.1 | 9, 10 | 1155 | |||
XM_017005559.1 | XP_016861048.1 | 9, 10 | 1182 | |||
XM_017005560.1 | XP_016861049.1 | 9, 10 | 1078 | |||
XM_017005561.1 | XP_016861050.1 | 9, 10 | 1183 | |||
XM_017005562.1 | XP_016861051.1 | 9, 10 | 1182 | |||
GenBank mRNA | AF117107.1 | - | 9, 10 | 1125 | ||
AK290664.1 | - | 9, 10 | 1159 | |||
BC021290.2 | - | 9, 10 | 1143 | 5090334 | ||
EU408701.1 | - | 8, 9 | 978 | |||
EU408702.1 | - | 8, 9 | 960 | |||
EU408704.1 | - | 7, 8 | 896 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
