siRNA Details
Gene Symbol: POM121L10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024593.1 | 5 | 6901 | |||
GenBank mRNA | BX648116.1 | - | 4 | 1993 |
Gene Symbol: LOC102725072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135922.1 | 5 | 2702 | |||
GenBank mRNA | BC051721.1 | - | 5 | 2702 |
Gene Symbol: LOC107987388 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030130.1 | XP_016885619.1 | 3 | 1807 | ||
XM_017030131.1 | XP_016885620.1 | 5 | 1260 | |||
XM_017030132.1 | XP_016885621.1 | 4 | 1280 |
Gene Symbol: POM121L8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024583.1 | 6 | 3763 | |||
GenBank mRNA | AL117485.1 | - | 4 | 1714 |
Gene Symbol: POM121L9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003714.1 | 6 | 2649 | |||
GenBank mRNA | AL040062.2 | - | 2 | 387 | ||
AL117401.1 | - | 6 | 2443 | |||
AL117484.1 | - | 3 | 845 | |||
BC063879.1 | - | 1 | 146 | |||
BC086868.1 | - | 2 | 971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
