siRNA Details
Gene Symbol: POM121L9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003714.1 | 6 | 2896 | |||
GenBank mRNA | AL117401.1 | - | 6 | 2690 | ||
BC063879.1 | - | 1 | 393 | |||
BC086868.1 | - | 2 | 1218 |
Gene Symbol: POM121L10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024593.1 | 5 | 7148 | |||
GenBank mRNA | BX648116.1 | - | 4 | 2240 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
