siRNA Details
Gene Symbol: POM121L9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003714.1 | 7 | 4876 | |||
GenBank mRNA | AL117401.1 | - | 7 | 4670 | ||
BX107726.1 | - | 1 | 66 |
Gene Symbol: LOC102725072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135922.1 | 6 | 4933 |
Gene Symbol: POM121L8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024583.1 | 7 | 5993 | |||
GenBank mRNA | BC034793.1 | - | 1 | 150 | ||
BC050342.1 | - | 1 | 420 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
