siRNA Details
Gene Symbol: IQSEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134382.2 | NP_001127854.1 | 4 | 1818 | ||
NM_014869.6 | NP_055684.3 | 4 | 2004 | |||
XM_011534304.2 | XP_011532606.1 | 6 | 2055 | |||
XM_011534305.2 | XP_011532607.1 | 4 | 1804 | |||
XM_011534306.2 | XP_011532608.1 | 4 | 2210 | |||
XM_011534307.2 | XP_011532609.1 | 5 | 2156 | |||
XM_011534308.2 | XP_011532610.1 | 4 | 1999 | |||
XM_011534310.2 | XP_011532612.1 | 6 | 2055 | |||
XM_011534311.2 | XP_011532613.1 | 4 | 2167 | |||
XM_011534312.2 | XP_011532614.1 | 4 | 1795 | |||
XM_011534313.2 | XP_011532615.1 | 4 | 2014 | |||
XM_011534314.2 | XP_011532616.1 | 4 | 1750 | |||
XM_011534315.2 | XP_011532617.1 | 4 | 1753 | |||
XM_017007581.1 | XP_016863070.1 | 6 | 2055 | |||
XM_017007582.1 | XP_016863071.1 | 5 | 2010 | |||
GenBank mRNA | AB018306.1 | - | 3 | 1497 | ||
AK294525.1 | - | 4 | 1818 | |||
AY653734.1 | - | 4 | 1766 | |||
BC010267.1 | - | 4 | 1758 | 2900367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
