siRNA Details
Gene Symbol: IQSEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134382.2 | NP_001127854.1 | 14 | 2953 | ||
XM_011534304.2 | XP_011532606.1 | 16 | 3190 | |||
XM_011534305.2 | XP_011532607.1 | 14 | 2939 | |||
XM_011534306.2 | XP_011532608.1 | 14 | 3345 | |||
XM_011534307.2 | XP_011532609.1 | 15 | 3291 | |||
XM_011534308.2 | XP_011532610.1 | 14 | 3134 | |||
XM_011534310.2 | XP_011532612.1 | 15 | 3140 | |||
XM_011534311.2 | XP_011532613.1 | 14 | 3302 | |||
XM_011534312.2 | XP_011532614.1 | 14 | 2930 | |||
XM_011534313.2 | XP_011532615.1 | 14 | 3149 | |||
XM_011534314.2 | XP_011532616.1 | 14 | 2885 | |||
XM_011534315.2 | XP_011532617.1 | 13 | 2838 | |||
XM_017007581.1 | XP_016863070.1 | 15 | 3160 | |||
XM_017007582.1 | XP_016863071.1 | 15 | 3145 | |||
GenBank mRNA | AK294525.1 | - | 14 | 2953 | ||
BC010267.1 | - | 13 | 2843 | 2900367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
