siRNA Details
Gene Symbol: RUFY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291994.1 | NP_001278923.1 | 14 | 2175 | ||
NM_014961.4 | NP_055776.1 | 13 | 2121 | |||
XM_011531755.2 | XP_011530057.1 | 13 | 1735 | |||
XM_017007891.1 | XP_016863380.1 | 14 | 1467 | |||
XM_017007892.1 | XP_016863381.1 | 13 | 1435 | |||
GenBank mRNA | AB020678.1 | - | 13 | 1927 | ||
AK294421.1 | - | 13 | 1306 | |||
AK296501.1 | - | 14 | 1362 | |||
BC051716.1 | - | 13 | 1989 | 5265230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
