siRNA Details
Gene Symbol: TTC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145418.1 | NP_001138890.1 | 22 | 5886 | ||
XM_005261405.2 | XP_005261462.1 | 21 | 5862 | |||
XM_006724171.3 | XP_006724234.1 | 21 | 14664 | |||
XM_011530018.2 | XP_011528320.1 | 22 | 6065 | |||
XM_017028673.1 | XP_016884162.1 | 22 | 5859 | |||
GenBank mRNA | AB028966.3 | - | 17 | 4679 | ||
AB665284.1 | - | 22 | 5834 | |||
BC016465.1 | - | 14 | 2430 | 4154050 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
