siRNA Details
Gene Symbol: TTC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145418.1 | NP_001138890.1 | 23 | 7955 | ||
XM_005261405.2 | XP_005261462.1 | 22 | 7931 | |||
XM_006724171.3 | XP_006724234.1 | 22 | 16733 | |||
XM_011530018.2 | XP_011528320.1 | 23 | 8134 | |||
XM_017028673.1 | XP_016884162.1 | 23 | 7928 | |||
GenBank mRNA | AB028966.3 | - | 18 | 6748 | ||
AB665284.1 | - | 23 | 7903 | |||
AL080174.1 | - | 2 | 2086 | |||
AL137328.1 | - | 1 | 723 | |||
BC016465.1 | - | 15 | 4499 | 4154050 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
