siRNA Details
Gene Symbol: WDPCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042692.2 | NP_001036157.1 | 7, 8 | 1458 | ||
NM_015910.5 | NP_056994.3 | 13, 14 | 2273 | |||
NR_122106.1 | 7, 8 | 1458 | ||||
XM_005264348.3 | XP_005264405.1 | 13, 14 | 2313 | |||
XM_011532881.2 | XP_011531183.1 | 14, 15 | 2097 | |||
XM_011532884.2 | XP_011531186.1 | 13, 14 | 2313 | |||
XM_011532887.2 | XP_011531189.1 | 13, 14 | 2313 | |||
XM_017004251.1 | XP_016859740.1 | 15, 16 | 2951 | |||
XM_017004252.1 | XP_016859741.1 | 14, 15 | 2097 | |||
GenBank mRNA | AF131737.1 | - | 7, 8 | 1458 | ||
AK022647.1 | - | 7, 8 | 1432 | |||
AL833623.1 | - | 13, 14 | 2059 | |||
BC093752.1 | - | 7, 8 | 1403 | 7939597 | ||
BC093754.1 | - | 7, 8 | 1403 | 7939599 | ||
BX647132.1 | - | 7, 8 | 4786 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
