siRNA Details
Gene Symbol: C3orf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171740.2 | NP_001165211.1 | 5 | 1004 | ||
NM_001171741.2 | NP_001165212.1 | 5 | 799 | |||
NM_001171743.2 | NP_001165214.2 | 5 | 783 | |||
NM_016210.4 | NP_057294.2 | 6 | 1093 | |||
XM_011533781.2 | XP_011532083.1 | 4 | 1743 | |||
XM_011533782.2 | XP_011532084.1 | 6 | 1081 | |||
XM_011533783.2 | XP_011532085.1 | 5 | 971 | |||
XM_011533784.2 | XP_011532086.1 | 6 | 859 | |||
XM_011533785.2 | XP_011532087.1 | 5 | 797 | |||
XM_011533788.1 | XP_011532090.1 | 4 | 403 | |||
XM_011533789.1 | XP_011532091.1 | 3 | 428 | |||
XM_011533790.1 | XP_011532092.1 | 6 | 1009 | |||
XM_017006545.1 | XP_016862034.1 | 5 | 921 | |||
XM_017006546.1 | XP_016862035.1 | 4 | 248 | |||
XM_017006547.1 | XP_016862036.1 | 5 | 944 | |||
GenBank mRNA | AF188706.1 | - | 5 | 764 | ||
AK124438.1 | - | 5 | 844 | |||
AK308074.1 | - | 5 | 665 | |||
AK309486.1 | - | 4 | 563 | |||
AK311093.1 | - | 2 | 284 | |||
AL833938.1 | - | 6 | 1052 | |||
BC034766.1 | - | 5 | 877 | 5177784 | ||
DA535127.1 | - | 4 | 439 | |||
DB027651.1 | - | 4 | 403 |
